missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate, 95%, Thermo Scientific™

805.78 DKK - 2560.00 DKK

Chemical Identifiers

CAS 67858-47-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD01571196
InChI Key GXSZDESAIZEFGZ-UHFFFAOYSA-N
Synonym methyl 4-formyl-1-methyl-1h-pyrrole-2-carboxylate, pubchem12439, methylformylmethylpyrrolecarboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-1-methyl-, methyl ester, 4-formyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester
PubChem CID 2798349
IUPAC Name methyl 4-formyl-1-methylpyrrole-2-carboxylate
SMILES CN1C=C(C=C1C(=O)OC)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10203672
View Documents Promotion Details
Thermo Scientific Maybridge
SEW06271CB
250 mg Amber Glass Bottle
805.78 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
10703843
View Documents Promotion Details
Thermo Scientific Maybridge
SEW06271DA
1 g Amber Glass Bottle
2560.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67858-47-3
167.164
GXSZDESAIZEFGZ-UHFFFAOYSA-N
2798349
CN1C=C(C=C1C(=O)OC)C=O
C8H9NO3
MFCD01571196
methyl 4-formyl-1-methyl-1h-pyrrole-2-carboxylate, pubchem12439, methylformylmethylpyrrolecarboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-1-methyl-, methyl ester, 4-formyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester
methyl 4-formyl-1-methylpyrrole-2-carboxylate

Specifications

67858-47-3
95%
MFCD01571196
GXSZDESAIZEFGZ-UHFFFAOYSA-N
methyl 4-formyl-1-methylpyrrole-2-carboxylate
2798349
95%
95.0
C8H9NO3
methyl 4-formyl-1-methyl-1h-pyrrole-2-carboxylate, pubchem12439, methylformylmethylpyrrolecarboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-1-methyl-, methyl ester, 4-formyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester
CN1C=C(C=C1C(=O)OC)C=O
167.164
167.16
Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications