missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylergometrine maleate, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
250 mg
Beschreibung
Active metabolite of methysergide (Cat. No. 1064); uterotonic agent and oxytocic.
Spezifikation
Spezifikation
Quantity | 50 mg |
Formula Weight | 455.51 |
Percent Purity | >98% |
Chemical Name or Material | Methylergometrine maleate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'