missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methysergide maleate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Mixed 5-HT1/5-HT2 receptor antagonist.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 469.54 |
Percent Purity | >99% |
Chemical Name or Material | Methysergide maleate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'