Learn More
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf Product Code.: 15927041
Estimated Shipment: 14-11-2025
Only null left
Add to basket
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf
Product Code. 15927041
Epoxy coated, 40.3 in., 10 lb., Each
Click to view available options
Finish:
Epoxy coated
Length (English):
28.3 in.
40.3 in.
Weight (English):
7.5 lb.
10 lb.
Unit Size:
Each
Product Code. 15927041
Brand: Metro™ LT48WE
Only null left
Estimated Shipment: 14-11-2025
Add to basket
Only null left
Add to basket
Expand your workspace in your lab. Metro Stainless Laboratory Worktable shelves come in a choice of stainless steel or epoxy-coated.
  • Shelves mount on three-sided frames with use of clips
  • Load rated for a maximum of 150 lbs.
  • Also compatible with MetroMax i Lab Worktables
Specifications
Product Type Wire Shelf
Finish Epoxy coated
For Use With (Equipment) Metro™ MetroMax i™ and other Stainless Steel worktables
Length (English) 40.3 in.
Length (Metric) 102.5 cm
Load Bearing Capacity (English) 150 lb.
Load Bearing Capacity (Metric) 68 kg
Mounting Type 3-Sided Frame
Weight (English) 10 lb.
Weight (Metric) 4.5 kg
Product Content Correction
Product Title
Metro™ Stainless Laboratory Worktable Accessory, Wire Shelf >