Learn More
Metro™ Super Adjustable™ Super Erecta™ Wire Shelf, Stainless Steel Finish Product Code.: 15906811
Only undefined left
Add to basket
Metro™ Super Adjustable™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Product Code. p-2387706
24 x 18 in. (61 x 45.7 cm), Each
Click to view available options
Dimensions (L x W):
24 x 14 in. (61 x 35.5 cm)
24 x 18 in. (61 x 45.7 cm)
24 x 2 in. (61 x 53 cm)
24 x 24 in. (61 x 61 cm)
30 x 14 in. (76.2 x 35.5 cm)
30 x 18 in. (76 x 45.7 cm)
30 x 2 in. (76.2 x 53 cm)
36 x 14 in. (91.4 x 35.5 cm)
36 x 18 in. (91.4 x 45.7 cm)
36 x 24 in. (91.4 x 61 cm)
36 x 30 in. (91.4 x 76.2 cm)
42 x 14 in. (106.6 x 35.5 cm)
42 x 18 in. (106.6 x 45.7 cm)
42 x 24 in. (106.6 x 61 cm)
48 x 14 in. (122 x 35.5 cm)
48 x 18 in. (121.9 x 45.7 cm)
48 x 24 in. (121.9 x 61 cm)
48 x 30 in. (122 x 76.2 cm)
54 x 18 in. (137 x 45.7 cm)
54 x 2 in. (137 x 53 cm)
54 x 24 in. (137 x 61 cm)
60 x 18 in. (152.4 x 45.7 cm)
60 x 24 in. (152.4 x 61 cm)
60 x 36 in. (152.4 x 91.4 cm)
72 x 18 in. (182.9 x 45.7 cm)
72 x 21 in. (182.9 x 53 cm)
72 x 24 in. (182.9 x 61 cm)
72 x 30 in. (183 x 76.2 cm)
72 x 36 in. (183 x 91.4 cm)
Unit Size:
Each
Product Code. 15906811
Brand: Metro™ A1824NS
Only undefined left
Add to basket
Only undefined left
Add to basket
Includes
One pack of nonconductive wedges (four pairs).
Requires
SiteSelect™ Posts (sold separately).
Wire shelf in stainless steel has a unique open-wire design permiting free circulation of air and minimizes dust accumulation.
  • Shelves adjust in 1 in. increments with patented shelf release corners
  • Shelf releases and wedges offer improved rigidity
  • Assembles quickly without tools
  • Assembled units are nonconductive
  • Maximum corrosion resistance
  • Nonporous surface is easily cleaned
  • Combine with Super Erecta Stationary or Mobile posts and accessories to create custom configurations for storage and transportation
  • Patented corner release system allows for easy height adjustment
  • Custom position your shelves for maximum storage density
  • Capacity: Shelves up to 48 in. wide; 800 lbs per shelf capacity (evenly distributed)
  • Shelves wider than 48 in.; 600 lbs per shelf (evenly distributed)
Warranty
One year
Specifications
Product Type Wire Shelf
Material Stainless Steel
Dimensions (L x W) 24 x 18 in. (61 x 45.7 cm)
For Use With (Equipment) Metro Super Erecta adjustable Shelving System
Includes Four non-conductive sleeve/wedges
Length (English) 24 in.
Length (Metric) 61 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Product Content Correction
Product Title
Metro™ Super Adjustable™ Super Erecta™ Wire Shelf, Stainless Steel Finish >