missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL20 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05952AF647
This item is not returnable.
View return policy
Description
METTL20 Polyclonal specifically detects METTL20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
METTL20 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C12orf72, chromosome 12 open reading frame 72, DKFZp451L235, EC 2.1.1.-, hypothetical protein LOC254013, methyltransferase like 20, MGC50559 | |
Produced in rabbits immunized with E. coli-derived Human METTL20 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
254013 | |
Store at 4°C in the dark. | |
IgG |