Learn More
Mettler Toledo™ Accessories for Seven2Go™ Conductivity Meters
For use with Seven2Go Conductivity meters.
Brand: Mettler Toledo™ 30122300
1245.98 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Seven2Go Conductivity Meters.
Specifications
uGo carrying case | |
Seven2Go Conductivity Meter | |
Carrying Case | |
uGo Carrying Case |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now
Product Content Correction
Product Title
Mettler Toledo™ Accessories for Seven2Go™ Conductivity Meters > uGo Carrying Case
Spot an opportunity for improvement?Share a Content Correction