Learn More
Mettler Toledo™ Accessories for Seven2Go™ Conductivity Meters
For use with Seven2Go Conductivity meters.
Brand: Mettler Toledo™ 30122303
136.16 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Seven2Go Conductivity Meters.
Specifications
Seven2Go meter benchtop stabilizing base | |
Bechtop Stabilizing Base | |
Benchtop Stabilizing Base |
Seven2Go Conductivity Meter | |
Providing extra stability on flat surfaces |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now
Product Content Correction
Product Title
Mettler Toledo™ Accessories for Seven2Go™ Conductivity Meters > Base Accessory
Spot an opportunity for improvement?Share a Content Correction