Learn More
Mettler Toledo™ Adapters for pH Meters
Accessory adapters
Brand:  Mettler Toledo™ 51302329
843.09 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11977731
  • 902.00 DKK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
In Stock
Description
LTW-MiniDin Conductivity Adapter
  • The appropriate cable to connect the conductivity sensor mini DIN to a portable meter.  Adapter is required for the following meters: FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LTW-MiniDin Conductivity Adapter
FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Mettler Toledo™ Adapters for pH Meters > Conductivity adapter