Learn More
Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
For use with ISE combination electrodes.
Brand: Mettler Toledo™ 51340029
244.35 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with ISE combination electrodes.
Specifications
Electrolyte for Bromide ISE, 20mL | |
ISE Filling Solution | |
Determine the Bromide concentration in samples | |
20 mL | |
0°C to 80°C |
Bromide ISE | |
1 mol/L KNO3 | |
Bottle | |
32°F to 176°F |
Product Content Correction
Product Title
Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions > Bromide Filling Solution
Spot an opportunity for improvement?Share a Content Correction