Learn More
Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters
The modular design of SevenExcellence allows you to expand your instrument with additional measurement parameters for future needs at any time.
Brand:  Mettler Toledo™ 30034471
9400.81 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13440689
  • 9645.00 DKK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
In Stock
Description
Just add the new expansion unit, switch on the instrument and the new expansion unit is automatically recognized and ready to use! SevenExcellence fits in total three expansion units in any composition.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±0.002
±0.1°C
-2 to 20
0.1/1mV
0.1°C
±0.1mV
pH/Ion Module
-2000 to 2000mV
0.001/0.01/0.1
-30°C to +130°C
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters > pH/Ion Module