Learn More
METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
4440.00 DKK - 27765.00 DKK
Product Code | Brand | Model | Channels | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Channels | Product Type | Price | Quantity & Availability | ||||
10205774
|
METTLER TOLEDO™
51302601 |
SevenGo Duo SG23 | 2 | Meter with Electrode Kit |
13160.00 DKK
1 set |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
10379304
|
METTLER TOLEDO™
51302600 |
SevenGo Duo SG23 | 2 | pH/Conductivity Meter |
8340.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
11967981
|
METTLER TOLEDO™
51302663 |
SevenGo Duo SG98 | 2 | Meter with Field Kit, 2m Cable |
25815.00 DKK
1 set |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
11918011
|
METTLER TOLEDO™
51302664 |
SevenGo Duo SG98 | 2 | Meter with Field Kit, 5m Cable |
27765.00 DKK
1 set |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
10422005
|
METTLER TOLEDO™
51302610 |
SevenGo Duo SG68 | 2 | pH/Ion/DO Meter |
15375.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
10717315
|
METTLER TOLEDO™
51302611 |
SevenGo Duo SG68 | 2 | Meter with Electrode Kit |
22950.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
10042274
|
METTLER TOLEDO™
51302621 |
SevenGo Duo SG78 | 2 | Meter with Electrode Kit |
17940.00 DKK
Each |
N/A | ||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
||||||||||
10297814
|
METTLER TOLEDO™
51302622 |
SevenGo Duo SG78 | 2 | Meter with Field Kit |
21070.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15345073
|
METTLER TOLEDO™
30207954 |
Seven2Go S3 | 1 | Conductivity Meter |
4940.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15355073
|
METTLER TOLEDO™
30207955 |
Seven2Go S3 | 1 | Meter with Standard Kit |
7005.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15365073
|
METTLER TOLEDO™
30207956 |
Seven2Go S3 | 1 | Meter with Field Kit |
7380.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
15375073
|
METTLER TOLEDO™
30207957 |
Seven2Go S3 | 1 | Meter with Bioethanol Kit |
7920.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
15385073
|
METTLER TOLEDO™
30207958 |
Seven2Go S4 | 1 | DO Meter |
5050.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15395073
|
METTLER TOLEDO™
30207959 |
Seven2Go S4 | 1 | Meter with Standard Kit |
8495.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15305083
|
METTLER TOLEDO™
30207960 |
Seven2Go S4 | 1 | Meter with Field Kit |
8990.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15315093
|
METTLER TOLEDO™
30207971 |
S9 DO Meter | 1 | Meter with Standard kit |
13545.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15325093
|
METTLER TOLEDO™
30207972 |
S9 DO Meter | 1 | Meter with Field kit |
13965.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15335093
|
METTLER TOLEDO™
30207939 |
S9 DO Meter | 1 | Meter with BOD Kit |
14460.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15395063
|
METTLER TOLEDO™
30207949 |
Seven2Go S2 | 1 | pH/mV Meter |
4440.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
15305073
|
METTLER TOLEDO™
30207950 |
Seven2Go S2 | 1 | Meter with Standard Kit |
5550.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15315073
|
METTLER TOLEDO™
30207951 |
Seven2Go S2 | 1 | Meter with Field Kit |
6010.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15325073
|
METTLER TOLEDO™
30207952 |
Seven2Go S2 | 1 | Meter with Food Kit |
6390.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
15335073
|
METTLER TOLEDO™
30207953 |
pH, ORP | 1 | Meter with Light Kit |
4900.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15355083
|
METTLER TOLEDO™
30207874 |
Seven2Go Pro S8 | 1 | pH/mV and Ion Meter |
8335.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15375083
|
METTLER TOLEDO™
30207877 |
Seven2Go Pro S8 | 1 | Meter with Field Kit |
8950.00 DKK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||
15385083
|
METTLER TOLEDO™
30207878 |
Seven2Go Pro S8 | 1 | Meter with Biotech Kit |
9720.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15395083
|
METTLER TOLEDO™
30207879 |
Seven2Go Pro S8 | 1 | Meter with Fluoride Kit |
12165.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
15305093
|
METTLER TOLEDO™
30207970 |
S9 DO Meter | 1 | DO Portable Meter |
7920.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
10430665
|
METTLER TOLEDO™
51302620 |
SevenGo Duo SG78 | 2 | pH/Ion/Cond Meter |
13005.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
11957981
|
METTLER TOLEDO™
51302661 |
SevenGo Duo SG98 | 2 | pH/Ion/DO/ORP Meter |
15565.00 DKK
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||
Description
- Dust proof
- Water proof
- IP67 Rated
- pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
- Compatible with a wide range of IP67-rated electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Data storage available; capacity dependent on model
- Export data to a printer or Excel file for Seven2Go Pro only
- EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
- Rubber holster available for single-handed measurements
- uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Specifications
ISO 9001:2000 | |
IP67 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters