Learn More
WTW™ MG 2.5 E Replacement Vessel
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Brand:  WTW™ 209512
185.78 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10064844
  • 195.00 DKK / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Specifications
2500 mL
Documents
One moment while we fetch your results.
Product Title
WTW™ MG 2.5 E Replacement Vessel > Capacity: 2500mL