missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICB Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHMICBX10
This item is not returnable.
View return policy
Description
MICB encodes the highly polymorphic MHC (HLA) class I chain-related gene B. The protein product is expressed on the cell surface. It is thought that MICB functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells. MICB is broadly recognized by NK cells and T cells with NKG2D receptor on their surface. The complex NKG2D-MICB results in MICB expressing cytolytic T cells and NK cells against epithelial tumor cells.Specifications
Q29980 | |
75 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
MICB | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.0686 to 50 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
4277 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
MICB | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction