Learn More
Fisherbrand™ Micro Insert for well plates, glass
1796.13 DKK valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Description
- Clear
- Glass
- Plates meet ANSI and SBS standards
- Different sealing options
- Chromatography tested
- No leeching due to high-end material
Specifications
Clear | |
Glass | |
1200 μL | |
Round |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70% -
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24163Expires: 2025-04-30
Product Content Correction
Product Title
Fisherbrand™ Micro Insert for well plates, glass > clear glass, 1200 μL Volume,U bottom
Spot an opportunity for improvement?Share a Content Correction