Learn More
Ansell Microgard™ 2500 Standard Overboots
Description
- Elastic to top of boot
- Tie fastening
- Bound seams
- Microporous polypropylene laminate
Specifications
Specifications
Material | Polypropylene |
Color | White |
Closure Type | Elastic Top with Ankle Ties |
Model | WH25-B-00-406-00 |
Seam Type | Bound |
Packaging Quantity | 200 Pairs |
Product Type | Boot Covers |