Description
- Microporous polypropylene laminate
- Fits Euro sizes 42 to 46 (UK 8 to 11)
- Elasticated opening
- ESD PVC anti-slip sole
- Adjustable Shoe Tie
- Bound seams
Specifications
Specifications
Material | Polypropylene |
Color | White |
Closure Type | Elastic with Adjustable Ties |
Sole Type | ESD PVC |
Model | WH25-B-00-407-00 |
Seam Type | Bound |
Packaging Quantity | 150 Pairs |
Product Type | Boot Covers |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Ansell Microgard™ 2500 Standard Overboots with ESD PVC Soles >
Spot an opportunity for improvement?Share a Content Correction