Learn More
Fisherbrand™ Micropipette Stand
Hold six pipettes in this classic stand. Fisherbrand™ Micropipettte Stand provides convenient, safe storage for most major pipette brands. Made with durable ABS in an attractive, solid blue stand.
Brand: Fisherbrand™ HS20613E
323.85 DKK valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Description
- Convenient and safe storage
- Classic six-place pipet stand designed to hold most major pipet brands
- Upper slots 25mm wide; Lower slots 20mm wide
Specifications
Micropipette Stand | |
Blue | |
6 |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
-
Multiple Manufacturers
Illuminating Protein Pathways
Shining a light on the tools that drive protein discovery.Promo Code: 24148Expires: 2025-08-22
New Offer Save Now -
Multiple Manufacturers
Sample Preparation, Filtration and Storage
Get up to 80% off some of our most popular products.Promo Code: 24124Expires: 2025-08-31 Redemption Only
New Offer Save Now