Learn More
Epredia™ Microscope Slides, Cut, 1mm
Shop All Epredia Products
Click to view available options
Dimensions (L x W):
45 x 30 mm
75 x 50 mm
76 x 26 mm
Quantity:
100 Pieces in Cardboard Box
100 Pieces in Paper
50 Pieces in Box
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
Description
- Manufactured from Swiss extra-white soda-lime glass
- Reduced iron content for low self-fluorescence
- Greenish shimmer minimized
- Guaranteed optimum, faultless transparency
- Clean, flat and suitable for immediate use
Specifications
Specifications
Length (Metric) | 75 mm |
Width (Metric) | 50 mm |
Material | Soda Lime Glass |
Thickness (Metric) | 1 mm |
Coating | Uncoated |
Quantity | 100 Pieces in Cardboard Box |
Writing Surface | No |
Dimensions (L x W) | 75 x 50 mm |
Product Content Correction
Product Title
Epredia™ Microscope Slides, Cut, 1mm >
Spot an opportunity for improvement?Share a Content Correction