Learn More
Merck Millicell™ Receiver Trays
Comes with lid
1070.00 DKK - 1100.00 DKK
Specifications
Closure Included | With Lid |
---|---|
Color | Clear |
Material | Polystyrene |
Shape | Rectangular |
Sterility | Sterile |
Product Code | Brand | Well Count | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Count | Price | Quantity & Availability | ||||||
10347753
|
Merck
PSSW010R5 |
1 |
1100.00 DKK
Pack of 5 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
10052523
|
Merck
PSMW010R5 |
24 |
1070.00 DKK
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
- 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
- Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
With Lid | |
Polystyrene | |
Sterile |
Clear | |
Rectangular | |
Receiver Tray |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Merck Millicell™ Receiver Trays