Learn More
Macherey-Nagel™ Mix V Diamino/Carbon Clean-up Mix
Brand: Macherey-Nagel™ 730975
This item is not returnable.
View return policy
Specifications
Chromabond QuEChERS Clean-up Mixes (EN 15662) | |
Diamino/Carbon Clean-up Mix | |
15 mL | |
50 Pack |
Product Content Correction
Product Title
Macherey-Nagel™ Mix V Diamino/Carbon Clean-up Mix > Volume: 15mL
Spot an opportunity for improvement?Share a Content Correction