Learn More
Köttermann Mobile Cabinet with One Lockable Door
This Mobile Cabinet with One Lockable Door offers versatile storage space with a single door to protect stored items.
Brand: Köttermann 315-00074
Description
- Versatile storage space
- Mobile with lockable wheels
- Features one door with locking mechanism to protect stored items
- Many sizes and configurations available
Specifications
White | |
516 x 600 x 740 mm | |
Epoxy Coated | |
Right | |
1 | |
None | |
Plastic with Brakes | |
Steel |
Mobile | |
Hinged | |
Steel | |
Yes | |
1 | |
None | |
Yes |