Learn More
Memmert™ MobileALERT
Notification by SMS with potential free contact error option
Brand:  Memmert™ IPP260ecoC3
 View more versions of this product
Product Code. 17170175
  • 9820.00 DKK / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Requires option H6
  • Notification by SMS in case of any error or alarm of the device
Specifications
Specifications
Requires option H6
IPP260eco
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ MobileALERT > IPP260eco