Learn More
Memmert™ MobileALERT For Up to 4 Alarm Notifications
MobileALERT for 2 alarm notifications, temperature and humidity
Brand:  Memmert™ ICO150C4
 View more versions of this product
Product Code. 17149954
  • 13255.00 DKK / Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Accessory
  • Notification by SMS, temperature and CO2 alarm standard
  • Additionally humidity alarm when equipped with option K7
  • O2 alarm when equipped with option T6
Specifications
Specifications
Temperature and humidity, up to 4 alarms
ICO150
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ MobileALERT For Up to 4 Alarm Notifications > ICO150