Learn More
Gibco™ Mock (G) SLC Transporter Cells
For use as a negative control in experiments utilizing TRANSiPORT solute carrier (SLC) transporter-overexpressing HEK293 cells
Brand: Gibco™ GM1101G
3709.75 DKK valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- These cells are intended for use as a negative control in experiments utilizing TRANSiPORT solute carrier (SLC) transporter-overexpressing HEK293 cells.
- These mock cells do not overexpress the SLC transport protein and so can be used to determine levels of endogenous transport activity as well as non-specific binding of compounds being tested.
Specifications
Store in liquid nitrogen vapor phase. | |
Mammalian Cell Culture | |
SLC Transporter Cell |
293 (HEK) | |
Human | |
8–10M cells |