Learn More
DuPont™ Tychem 6000 F CHA5 Hooded Coverall
GREENER_CHOICE
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
Brand:  DuPont™ D13395545
6680.19 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11703093
  • 7165.00 DKK / Pack of 25
Only null left
Estimated Shipment: 15-08-2025
Description
  • Available in grey for discretionary needs
  • Elasticated face, wrists, waist and ankles for optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive chin flap for tight seal of suit to face masks
  • Self-adhesive double zipper flap closure system
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hooded Coveralls
Tychem
Elastic
Elastic
Elastic
Stitched, Taped
Chemicals
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
X-Large
Gray
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
Self-adhesive chin flap
108 to 116 cm
Documents
Product Information
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
DuPont™ Tychem 6000 F CHA5 Hooded Coverall > Size: X-Large