missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde)
Click to view available options
Quantity:
10 mg
Unit Size:
Each
Description
- High sensitivity, freedom from background and long-wavelength excitability make these potential reagents for researcher, diagnostic and forensic applications
- Resulting products of CBQCA are maximally excited at 450nm or by 442nm spectral line of He-Cd laser
- Have emission maxima at approx. 550nm
- In capillary zone electrophoresis (CZE), sensitivity of detection of ATTO-TAG CBQCA conjugates should be in attomole range (10-18 moles)
Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Amine |
Reactive Moiety | Aldehyde |
Shipping Condition | Room Temperature |
Form | Solid |
Product Line | ATTO-TAG™ |
Quantity | 10 mg |
For Use With (Application) | Cell Viability, Proliferation and Function; Protein Electrophoresis and Western Blotting |
Target | Primary Amine |
Type | Derivatization Kits and Reagents |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde) >
Spot an opportunity for improvement?Share a Content Correction