Learn More
Invitrogen™ BackDrop™ Background Suppressor for Microplate Assays
Designed to effectively suppress background fluorescence in live cell assays
Brand:  Invitrogen™ B10511
 View more versions of this product
Product Code. 15421094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
BackDrop Background Suppressor is a novel signal enhancer reagent that helps improve signal-to-noise ratios when used with green-emitting fluorescent dyes or proteins.
  • Can be added to complete media
  • Improves sensitivity of green-emitting fluorescent dyes or proteins
  • Validated with many cellular assays, including ion indicator and membrane potential assays
  • Minimal cell toxicity—demonstrated to not cause osmotic shock to neuronal cells
The sensitivity of fluorescent assays can be compromised by autofluorescence or background fluorescent signals. These background signals result from cellular proteins or growth media components and contribute to a decrease in the signal-to-noise ratio and reduced assay sensitivity. Addition of BackDrop Background Suppressor significantly reduce background signal, helping improve experimental results. BackDrop Background Suppressor can be added to growth media and has been formulated to not contribute to cell toxicity. It has been successfully used with many different cell types and fluorescent reagents (including ion indicators) to efficiently suppress background fluorescence.
Cell Analysis, Cellular Imaging, Immunocytochemistry (ICC)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
One 5 mL bottle
Store at 2-8°C. DO NOT FREEZE. Protect material from long-term exposure to light.
Microplate Assays
5 mL
BackDrop™
Background Suppressor for live cell assays
Documents
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2031050 Certificate TypeCertificate of Analysis Date04/09/2018 Catalog NumberB10511
Lot Number1968984 Certificate TypeCertificate of Analysis Date19/03/2018 Catalog NumberB10511
Lot Number1907870 Certificate TypeCertificate of Analysis Date02/08/2017 Catalog NumberB10511
Lot Number1633498 Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberB10511
Lot Number1773118 Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberB10511
Product Title
Invitrogen™ BackDrop™ Background Suppressor for Microplate Assays > 5mL