Learn More
Invitrogen™ BODIPY™ FL C12-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl) Sphingosyl Phosphocholine) Product Code.: 10073292
Invitrogen™ BODIPY™ FL C12-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl) Sphingosyl Phosphocholine)
Product Code. 10073292
250 μL, Each
Click to view available options
Quantity:
250 μL
Unit Size:
Each
Product Code. 10073292
Brand: Invitrogen™ D7711
Green-fluorescent sphingomyelin
BODIPY FL C12-sphingomyelin can be used to study sphingolipid transport and signaling pathway in cells.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sphingolipids
Quantity 250 μL
Product Line BODIPY
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2916322 Certificate TypeCertificate of Analysis Date22/08/2024 Product CodeD7711
Lot Number2806859 Certificate TypeCertificate of Analysis Date30/12/2023 Product CodeD7711
Lot Number2626233 Certificate TypeCertificate of Analysis Date02/08/2023 Product CodeD7711
Lot Number2456916 Certificate TypeCertificate of Analysis Date09/03/2022 Product CodeD7711
Lot Number2301034 Certificate TypeCertificate of Analysis Date31/03/2021 Product CodeD7711
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ BODIPY™ FL C12-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl) Sphingosyl Phosphocholine) >