Learn More
Invitrogen™ BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine) Product Code.: 11510306
Invitrogen™ BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine)
Product Code. 11510306
250 μg, 250µg
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
Product Code. 11510306
Brand: Invitrogen™ D3522
Useful in cellular imaging
BODIPY FL C5 sphingomyelin can be used in sphingolipid transport and metabolism studies.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sphingolipids
Quantity 250 μg
Physical Form Solid
Product Line BODIPY
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number3061372 Certificate TypeCertificate of Analysis Date09/10/2024 Product CodeD3522
Lot Number2971120 Certificate TypeCertificate of Analysis Date04/06/2024 Product CodeD3522
Lot Number2785495 Certificate TypeCertificate of Analysis Date30/08/2023 Product CodeD3522
Lot Number2448078 Certificate TypeCertificate of Analysis Date26/01/2022 Product CodeD3522
Lot Number2281175 Certificate TypeCertificate of Analysis Date21/12/2020 Product CodeD3522
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine) >