Description
BODIPY TR ceramide can be used to stain Golgi apparatus in live cells.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Chemical Name or Material | Sphingolipids |
Quantity | 250 μg |
Physical Form | Solid |
Product Line | BODIPY |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ BODIPY™ TR Ceramide >
Spot an opportunity for improvement?Share a Content Correction