Learn More
Invitrogen™ BODIPY™ TR Ceramide complexed to BSA
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The red-fluorescent BODIPY TR C5 ceramide complexed to bovine serum albumin. Complexing fluorescent lipids with BSA facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe; the BSA-complexed probe can be directly dissolved in water.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Chemical Name or Material | Sphingolipids |
Quantity | 5 mg |
Physical Form | Solid |
Product Line | BODIPY |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ BODIPY™ TR Ceramide complexed to BSA >
Spot an opportunity for improvement?Share a Content Correction