Learn More
Invitrogen™ Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE) Product Code.: 10154432
Invitrogen™ Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE)
Product Code. 10154432
1 mg, Each
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Product Code. 10154432
Brand: Invitrogen™ O12650
With excitation/emission maxima ∽501/526 nm
The phospholipid, Oregon Green 488 DHPE is labeled on the head group with the green-fluorescent Oregon Green fluorophore. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∽4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
Cell Analysis, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, pH Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phospholipids
Quantity 1 mg
Physical Form Solid
Product Line Oregon Green
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number3080061 Certificate TypeCertificate of Analysis Date21/10/2024 Product CodeO12650
Lot Number3001518 Certificate TypeCertificate of Analysis Date03/08/2024 Product CodeO12650
Lot Number2980670 Certificate TypeCertificate of Analysis Date28/06/2024 Product CodeO12650
Lot Number2961113 Certificate TypeCertificate of Analysis Date03/06/2024 Product CodeO12650
Lot Number2528047 Certificate TypeCertificate of Analysis Date01/09/2022 Product CodeO12650
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE) >