missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
30 mL
125 mL
250 mL
500 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mo | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Packaging | Poly Bottle |
CAS Min % | 0.1, 99.9 |
Quantity | 250 mL |
CAS Max % | 0.1, 99.9 |
Certificate of Analysis | Assurance Certificate of Analysis reports up to 30 elements at ppb levels. |
Technique | AA, ICP |
Type | Certified Reference Material |
Grade | Assurance |
Chemical Name or Material | Molybdenum Standard |
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molybdenum Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction