Learn More
Invitrogen™ Mouse Thyroid Stimulating Hormone ELISA Kit
For the quantitative detection of Mouse Thyroid Stimulating
Brand:  Invitrogen™ EEL110
Product Code. 18007374
  • 4594.00 DKK / Pack of 96
Explore more special offers
Description
Description
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P01216, P12656
0.75 ng/mL
ELISA
Colorimetric Microplate Reader
CGA, TSHB
<10%
HRP
RUO
Refer to product documentation for component specific storage temperature.
1 hr 20 min
1.25 to 80 ng/mL
HRP
Plasma
12640, 22094
<10%
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
96 Tests
100 µL
Mouse
3 hr 30 min