Learn More
Invitrogen™ Mouse Thyroid Stimulating Hormone ELISA Kit
For the quantitative detection of Mouse Thyroid Stimulating
Brand: Invitrogen™ EEL110
This item is not returnable.
View return policy
Description
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.Specifications
P01216, P12656 | |
0.75 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CGA, TSHB | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
1.25 to 80 ng/mL | |
HRP | |
Plasma | |
12640, 22094 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
100 µL | |
Mouse | |
3 hr 30 min |