Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Magnesium and Calcium
Shop All MP Biomedical Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Calcium and magnesium support cell adhesion. Hanks' Balanced Salts (Modified), without Magnesium and Calcium, is used to minimize cell attachment or aggregation.
Specifications
Specifications
Color | Red |
Content And Storage | 15°to 30°C |
Dilution | 1 X |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Quantity | 500 mL |
Formulation | Potassium Chloride 400 mg/L, Potassium Phosphate Monobasic 60 mg/L, Sodium Bicarbonate 350 mg/L, Sodium Chloride 8000 mg/L, Sodium Phosphate Dibasic 47.50 mg/L, Dextrose 1000 mg/L, Phenol Red.Sodium Salt 17 mg/L |
pH | 6.9 to 7.4 |
Form | Liquid |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Product Title
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Magnesium and Calcium >
Spot an opportunity for improvement?Share a Content Correction