missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ L-Broth, Powder
Luria-Broth, Rich Medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium
Brand: MP Biomedicals™ 113001032
This item is not returnable.
View return policy
Description
Use 16 grams per liter
Contents/liter: 10g tryptone, 5g yeast extract, 0.5g NaCl
Specifications
15° to 30°C | |
Sterile | |
1 kg |
Bacterial Growth Medium | |
6.9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction