Learn More
MP Biomedicals™ MP EcoChrom Polyamide (for TLC)
MP EcoChrom Polyamide (for TLC)
Brand:  MP Biomedicals™ 209603
Product Code. 30058482
  • 1750.00 DKK / 250g
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 to 20 μm
Polyamide
TLC
Chromatography
Polyamide
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ MP EcoChrom Polyamide (for TLC) > Particle Size: 5 to 20 μm