Learn More
Plum™ MP2000 Dispenser
Flexible dispenser system for bag-in-box products (soap/ hand cleaning/ hand lotion). Well-suited for production plants, repair shops, food processing, technical colleges and graphical industries.
Brand:  Plum™ P4251
 View more versions of this product
Product Code. 15761137
  • 766.00 DKK / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Description
  • Hygienic and reliable dispenser design
  • Easily customised to hand hygiene requirements in the individual workplace
  • MP2000 products are supplied in airtight packages for prevention of bacteria growth
  • Easily mounted, cleaned and refilled
  • Specifications
    Specifications
    110 mm
    130 mm
    300 mm
    1
    Product Title
    Plum™ MP2000 Dispenser > Width (Metric): 130 mm