Learn More
MP Biomedicals MPure-32 aNAP System
This is a fully automated magnetic bead-based nucleic acid extraction system. It can process up to 32 samples simultaneously within a short period of time (around 40 to 60 minutes).
Brand: MP Biomedicals 07EMC043
Description
MPure-32™ aNAP System is a fully automated magnetic bead-based nucleic acid extraction system. It can process up to 32 samples simultaneously within a short period of time (around 40 to 60 minutes). Besides its capability to extract nucleic acid with high purity and yield, MPure-32™ aNAP System is easy to use; this eliminates the risk of human error and cross-contamination.
- Reduces working time
- Minimal contamination risk
- Superior Purity and Yield
- High Flexibility (Open to Customization)
- Simple Workflow
- Fast and Consistent Results
- Run Time : 45 to 60 minutes
- Samples per run : From 1 to 3 samples
- Weight (NW) : 21 kg
- Dimensions : 38 cm x 36 cm x 37 cm
- Power Supply : 3.2 A, 100/240 V
- Processing Volume : 50 ÎĽL to 1000 ÎĽL
- Magnetic Rod : >4,300 gauss
- Temp Control : 1 set
- Heating Block : Yes (2 pcs)
- Heating : RT - 70°C
- UV Lamp : Yes
- Display : 5.5 in. Touch Screen
MP Biomedicals also offers a range of reagent kits for various types of biological samples. The user has the flexibility to self-assemble the reagents with the relevant consumables for maximum cost-effectiveness or purchase the pre-assembled reagent kits for greater convenience.

Specifications
Viruses, blood, plant and animal cells, feces, soil samples, bacteria, yeast, saliva |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Let s get to know MP Biomedicals MPure-32 aNAP System
Your input is important to us. Please complete this form to provide feedback related to the content on this product.