Learn More
Tocris Bioscience™ MRS 2578
Selective P2Y6 antagonist
1265.00 DKK - 5005.00 DKK
Specifications
Product Type | MRS 2578 |
---|---|
Molecular Weight (g/mol) | 472.66 |
Inhibitors | MRS 2578 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745586
|
Tocris Bioscience™
2146/10 |
10 mg |
1265.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18004492
|
Tocris Bioscience™
2146/50 |
50 mg |
5005.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.Specifications
MRS 2578 | |
472.66 | |
MRS 2578 |
For Research Use Only