Learn More
Eppendorf™ MTP Flex Centrifuge Bucket
Use these buckets with Eppendorf™ Rotor A-4-81.
Brand: Eppendorf™ 5810742005
6123.40 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- MTP Flex bucket
- Available in packs of two and four
- Compatible with Rotor A-4-81
Specifications
Eppendorf A-4-81-MTP retrofit set | |
2/Pk. |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30