missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Mueller Hinton Agar with Horse Blood
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium for fastidious organisms according to EUCAST.
Brand: Thermo Scientific™ PB5303A
120.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar with Horse Blood, a medium designed and manufactured according to the European standard, EUCAST.
The addition of horse blood and NAD enable the growth of fastidious organisms and the formulation ensures that toxic factors will be absorbed to establish better growth from very small inocula. Conforms to EUCAST.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
According to EUCAST | |
2° to 12°C, in the dark | |
Germany | |
Agar | |
Flow Wrapped - Single Layer |
Signal red, opaque | |
90mm Monoplate | |
MHA with Horse Blood | |
24g ± 5% | |
10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction