Learn More
Thermo Scientific™ Muller-Kauffmann Tetrathionate Broth
Selectively enrich for Salmonella species with the convenience of ready prepared media tubes.
Brand: Thermo Scientific™ TV5006I
587.11 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Selectively enrich for Salmonella species from food samples with Thermo Scientific™ Muller-Kauffmann Tetrathionate Broth in convenient ready prepared media tubes. Muller1 developed this medium in 1923. It was later modified by Kauffmann2,3 with the addition of brilliant green and ox bile to suppress commensal organisms and, thus, improve the isolation of salmonellae.
The brilliant green dye used in Muller-Kauffmann Tetrathionate Broth has been shown to be critical. Incubation of Muller-Kauffmann Tetrathionate Broth at 43°C was also shown to be essential.
The technique used for enrichment of the salmonellae is as follows:
Add approximately 10g of sample to 100mL of Muller-Kauffmann Tetrathionate Broth. Shake vigorously and immediately place the flasks of medium in a 45°C water-bath for 15 minutes. Remove the flasks from the water-bath, without drying them, and place in an incubator or another water-bath at 43°C.
Sub-culture the broth after 18 to 24 hours and again after 48 hours. Take one loopful of broth from the edge of
the surface of the fluid and inoculate either two Thermo Scientific Brilliant Green Agar (Part No. PO5033A) without recharging the loop between plates. Incubate the plates at 35°C for 18 to 24 hours.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Light green, opaque | |
Muller-Kauffmann Tetrathionate Broth | |
Germany | |
10 mL | |
50 tubes in a carton box |
2°C to 12°C | |
Tube | |
Microbiology Media | |
10 ± 0.5mL | |
50/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6003896 | Certificate of Analysis | 05/04/2024 | TV5006I |
3811040 | Certificate of Analysis | 23/03/2024 | TV5006I |
3798296 | Certificate of Analysis | 14/02/2024 | TV5006I |
3792456 | Certificate of Analysis | 30/01/2024 | TV5006I |
3777114 | Certificate of Analysis | 28/12/2023 | TV5006I |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For In Vitro Diagnostic Use.