Learn More
Heidolph™ Multi-channel Pump Head
Increase your throughput by running more cassettes on one single pump drive and pump head system
8095.00 DKK - 33210.00 DKK
Product Code | Brand | Model | Holds | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Holds | Includes | Price | Quantity & Availability | |||
10448401
|
Heidolph™
524-80420-00 |
C 4 | 4 small cassettes | 8 rollers |
8095.00 DKK
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||
10504742
|
Heidolph™
524-40810-00 |
C 8 | 8 medium cassettes | 4 rollers |
11790.00 DKK
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
10504932
|
Heidolph™
524-81220-00 |
C 12 | 12 small cassettes | 8 rollers |
33210.00 DKK
Each |
|
|||
Description
- Separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rate
- Save time - tubing change is done in just seconds
- Change your cassettes easily, even during operation – there are no restrictions
- Cassette adjustments and changes are simple, just click in place
- No additional purchase of a pump drive needed: upgrade your single-channel pump drive with just one adaptor for multi-channel use and pick the head/cassette configuration that matches your application needs
Specifications
Pump Head |