Learn More
Fisherbrand™ Flexi Caster
Accessory for use with Sub-Gel Midi-Plus or Maxi Horizontal Gel Electrophoresis System.
Brand:  Fisherbrand™ MS15/20-FC
Product Code. 11813363
  • 1420.00 DKK / Each
Estimated Shipment: 11-06-2025
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with Sub-Gel Midi-Plus or Maxi Horizontal Gel Electrophoresis System
Specifications
Specifications
Sub-Gel Midi-Plus or Maxi Horizontal Gel Electrophoresis System
Flexi Caster
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand and Fisher BioReagents Focus on electrophoresis
    Product Title
    Fisherbrand™ Flexi Caster > Gel Dimensions (LxW): Adjustable sizes: 150 x 70, 100, and 150mm, 200 x 100, 150, and 200mm, and 200 x 250mm