Learn More
Thermo Scientific™ Power Adapters for Digital Vortex Mixers
Power adapters for Digital Vortex Mixers
433.40 DKK
Specifications
Material Plastic, cable, rubber, screw
Color Black
For Use With (Equipment) Digital Vortex Mixer
Includes Power adapter
Packaging Quantity 1 Ea.
View More Specs
Products 1
Product Code Brand Description Plug Type Price Quantity & Availability  
Product Code Brand Description Plug Type Price Quantity & Availability  
16338816
View Documents
Thermo Scientific™
88870124
Multi-power adapter 220V EU, UK, CN Plug
433.40 DKK
Each
In Stock
Add to basket
Description
Description
Power adapters for Digital Vortex Mixers, which offer accurate and efficient sample mixing
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Warranty
2 years
Specifications
Specifications
Plastic, cable, rubber, screw
Digital Vortex Mixer
1 Ea.
Black
Power adapter
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Power Adapters for Digital Vortex Mixers