missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Myosin Heavy Chain 3 Antibody (3H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00004621-M06
This item is not returnable.
View return policy
Description
Myosin Heavy Chain 3 Monoclonal antibody specifically detects Myosin Heavy Chain 3 in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
Myosin Heavy Chain 3 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
HEMHC, Muscle embryonic myosin heavy chain, MYHC-EMB, MYHSE1, Myosin heavy chain, fast skeletal muscle, embryonic, myosin, heavy chain 3, skeletal muscle, embryonic, myosin, heavy polypeptide 3, skeletal muscle, embryonic, myosin, skeletal, heavy chain, embryonic 1, myosin-3, SMHCEMyosin heavy chain 3 | |
MYH3 (NP_002461.1, 2 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SSDTEMEVFGIAAPFLRKSEKERIEAQNQPFDAKTYCFVVDSKEEYAKGKIKSSQDGKVTVETEDNRTLVVKPEDVYAMNPPKFDRIEDMAMLTHLNEP | |
0.1 mg | |
Dendritic Cell Markers, Immunology | |
4621 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Sandwich ELISA | |
3H3 | |
Western Blot 1:500, ELISA, Sandwich ELISA | |
NP_002461 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |