missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-Triacetic Acid Trisodium Salt Solution, Honeywell™ Fluka™

Product Code. 15695810
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

for complexometry,∼35% in H2O (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 139-89-9
Quantity 1 L
Molecular Formula C10H15N2Na3O7
Linear Formula HOCH2CH2N(CH2COONa)CH2CH2N(CH2COONa)2
MDL Number MFCD00043341
UN Number NONH for all modes of transport
Synonym trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3
InChI Key WHNXAQZPEBNFBC-UHFFFAOYSA-K
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate
Molecular Weight (g/mol) 344.206
PubChem CID 8772
Formula Weight 344.2g/mol
Packaging Glass bottle
Chemical Name or Material N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid trisodium salt solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.