Learn More
N(alpha)-Boc-L-glutamine, 98+%, Thermo Scientific™
229.00 DKK - 741.00 DKK
Chemical Identifiers
CAS | 13726-85-7 |
---|---|
Molecular Formula | C10H17N2O5 |
Molecular Weight (g/mol) | 245.26 |
MDL Number | MFCD00065571 |
InChI Key | VVNYDCGZZSTUBC-LURJTMIESA-M |
Synonym | boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamineShow More |
PubChem CID | 83687 |
IUPAC Name | (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed in the synthesis and analgesic activity of human .beta.-endorphin.
Solubility
Soluble in DMSO and methanol. Soluble in 1 mmole in 2 ml DMF.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
13726-85-7 | |
245.26 | |
VVNYDCGZZSTUBC-LURJTMIESA-M | |
83687 | |
CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O |
C10H17N2O5 | |
MFCD00065571 | |
boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamineShow More | |
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acidShow More |
Specifications
118°C to 120°C | |
C10H17N2O5 | |
2127805 | |
Soluble in DMSO and methanol. Soluble in 1?mmole in 2 ml DMF. | |
CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O | |
245.26 | |
246.26 | |
≥98% |
13726-85-7 | |
MFCD00065571 | |
boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine | |
VVNYDCGZZSTUBC-LURJTMIESA-M | |
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid | |
83687 | |
−18.5° (c=1, DMF) | |
N(alpha)-Boc-L-glutamine |
Safety and Handling
missing translation for 'einecsNumber' : 237-296-8
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
Your input is important to us. Please complete this form to provide feedback related to the content on this product.